EMAIL THIS PAGE TO A FRIEND
European journal of biochemistry
Histone phosphorylation and chromatin structure during mitosis in Chinese hamster cells.
PMID 206429
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
369A-1
Phosphohistone H3 (PHH3) Rabbit Polyclonal Antibody