EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
Facilitation of Ih channels by P2Y1 receptors activation in Mesencephalic trigeminal neurons.
PMID 20643189
Abstract
P2Y(1) receptors, a subset of G-protein coupled receptors, have been shown to participate in sensory transduction in the periphery nervous system. However, little is known about their sensory function in the central nervous system. Here, by using immunohistochemistry, we showed that P2Y(1) receptors are predominantly localized in the somata of Mesencephalic trigeminal neurons (Mes V neurons), the primary sensory neurons in brainstem. Whole-cell voltage-clamp recording revealed that ADP-beta-S, a P2Y receptor agonist, enhanced the activity of hyperpolarization-activated cation channels (Ih channels) in Mes V neurons and that the activity-enhancing effect of ADP-beta-S could be blocked by a specific P2Y(1) receptor antagonist, MRS 2179. Taken together, these results suggested a possible role of P2Y(1) receptors in the information transduction of central sensory neurons through regulating Ih channel activities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8016
Adenosine 5′-[β-thio]diphosphate trilithium salt, ≥80% (HPLC)
C10H12Li3N5O9P2S