EMAIL THIS PAGE TO A FRIEND
Journal of veterinary pharmacology and therapeutics
Insect nicotinic acetylcholine receptor agonists as flea adulticides in small animals.
PMID 20646191
Abstract
Fleas are significant ectoparasites of small animals. They can be a severe irritant to animals and serve as a vector for a number of infectious diseases. In this article, we discuss the pharmacological characteristics of four insect nicotinic acetylcholine receptor (nAChR) agonists used as flea adulticides in dogs and cats, which include three neonicotinoids (imidacloprid, nitenpyram, and dinotefuran) and a macrocyclic lactone (spinosad). Insect nAChR agonists are one of the most important classes of insecticides, which are used to control sucking insects on both plants and animals. These novel compounds provide a new approach for practitioners to safely and effectively eliminate adult fleas.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32499
Dinotefuran, PESTANAL®, analytical standard
C7H14N4O3