EMAIL THIS PAGE TO A FRIEND
Nanoscale
Fabrication and characterization of polymer insulated carbon nanotube modified electrochemical nanoprobes.
PMID 20648318
Abstract
Electrochemical nanoprobes were fabricated from polymer insulated multiwalled carbon nanotube modified tapping mode atomic force microscope probes. An electrochemically active length of carbon nanotube was exposed by laser ablation of the insulating polymer. Characterization of these probes is done by cyclic voltammetry of ferrocenemethanol in an aqueous solution and by finite element analysis. The fabricated nanoelectrodes were found to be stable and yielded an interfacial electron transfer rate constant (k(0)) of 1.073 +/- 0.36 cm s(-1) for ferrocenemethanol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
335061
Ferrocenemethanol, 97%
C11H12FeO