EMAIL THIS PAGE TO A FRIEND
Journal of dental research
Substance P sensitizes P2X3 in nociceptive trigeminal neurons.
PMID 20651096
Abstract
Peripheral inflammation produces pain hypersensitivity by sensitizing nociceptors. Potentiation of P2X3 receptor activity in nociceptors may play an important role in this peripheral sensitization. However, we do not fully understand how P2X3 activity is elevated in inflammation. Thus, we investigated whether P2X3 activity in trigeminal nociceptive neurons is regulated by the neurokinin-1 (NK-1) receptor that is activated by an inflammatory mediator, substance P. Single-cell RT-PCR and immunohistochemistry revealed that NK-1 in nociceptive neurons was mainly co-expressed with P2X3. Ca(2+) imaging and whole-cell patch-clamp recordings indicated that both substance P and Sar-substance P, a selective NK-1 agonist, significantly potentiated α,β-meATP-induced currents and [Ca(2+)](i) responses in nociceptive neurons. These potentiating effects were completely blocked by GR82334, a specific NK-1 antagonist. Our results demonstrate that substance P sensitizes P2X3 receptor through the activation of NK-1, thus warranting these receptors as possible targets for pain therapy in the orofacial region. α,β-methylene adenosine 5'-triphosphate (ATP), α,β-meATP; neurokinin-1, NK-1; single-cell reverse-transcription polymerase chain-reaction, single-cell RT-PCR; [Sar(9),Met(O(2))(11)]-substance P, Sar-substance P.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S3672
[Sar9, Met(O2)11]-Substance P, ≥95% (HPLC)
C64H100N18O15S