EMAIL THIS PAGE TO A FRIEND
Journal of nutritional science and vitaminology
Development of simultaneous enzymatic assay method for all six individual vitamin B6 forms and pyridoxine-beta-glucoside.
PMID 20651455
Abstract
A method for determining all of the six natural vitamin B(6) compounds and pyridoxine-beta-glucoside in urine from humans consuming their usual diet was developed. These compounds were specifically converted with 5 enzymes into a high fluorescent 4-pyridoxolactone which was supersensitively determined by an isocratic HPLC. All of the compounds in urine from humans consuming their usual diets were for the first time determined together. The preparation procedure for urine samples was easy without HCl-hydrolysis, and the enzyme reactions took only 2 or 3 h. It required only 5 microL of the urine sample for analysis of one of the compounds. Urine samples from five young Japanese males consuming their usual diet contained pyridoxal, pyridoxamine, and pyridoxine-beta-glucoside but not pyridoxine or phosphoester forms. The contents of 4-pyridoxic acid and pyridoxal correlate well with a correlation coefficient of 0.98. On the other hand, the content of pyridoxamine did not correlate with that of 4-pyridoxic acid.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P9630
4-Pyridoxic acid, ≥98%
C8H9NO4