EMAIL THIS PAGE TO A FRIEND
Journal of nutritional science and vitaminology
Difructose anhydride III enhances bioavailability of water-insoluble iron in anemic Vietnamese women.
PMID 20651460
Abstract
Difructose anhydride III (DFAIII) is an indigestible disaccharide and has been shown to enhance iron absorption in animal studies; however, the effect has not been investigated in anemic subjects. We investigated the efficacy of co-administration of DFAIII with water-insoluble iron in the treatment of iron deficiency anemia in Vietnamese women. One hundred sixty-eight moderately anemic women (80 g/L
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P6526
Iron(III) pyrophosphate, soluble crystals
Fe4O21P6