EMAIL THIS PAGE TO A FRIEND
Experimental lung research
Pretreatment with perfluorohexane vapor attenuates fMLP-induced lung injury in isolated perfused rabbit lungs.
PMID 20653469
Abstract
The authors investigated the protective effects and dose dependency of perfluorohexane (PFH) vapor on leukocyte-mediated lung injury in isolated, perfused, and ventilated rabbit lungs. Lungs received either 18 vol.% (n = 7), 9 vol.% (n = 7), or 4.5 vol.% (n = 7) PFH. Fifteen minutes after beginning of PFH application, lung injury was induced with formyl-Met-Leu-Phe (fMLP). Control lungs (n = 7) received fMLP only. In addition 5 lungs (PFH-sham) remained uninjured receiving 18 vol.% PFH only. Pulmonary artery pressure (mPAP), peak inspiratory pressure (P(max)), and lung weight were monitored for 90 minutes. Perfusate samples were taken at regular intervals for analysis and representative lungs were fixed for histological analysis. In the control, fMLP application led to a significant increase of mPAP, P(max), lung weight, and lipid mediators. Pretreatment with PFH attenuated the rise in these parameters. This was accompanied by preservation of the structural integrity of the alveolar architecture and air-blood barrier. In uninjured lungs, mPAP, P(max), lung weight, and lipid mediator formation remained uneffected in the presence of PFH. The authors concluded that pretreatment with PFH vapor leads to an attenuation of leukocyte-mediated lung injury. Vaporization of perfluorocarbons (PFCs) offers new therapeutic options, making use of their protective and anti-inflammatory properties in prophylaxis or in early treatment of acute lung injury.
Related Materials