EMAIL THIS PAGE TO A FRIEND
Histopathology
Risk factors in thyroid-type carcinoma arising in ovarian struma: a report of 15 cases with comparison to ordinary struma ovarii.
PMID 20653787
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05-954
Anti-TCL1 Antibody, culture supernatant, clone, Upstate®