EMAIL THIS PAGE TO A FRIEND
FEBS letters
The 'true' L-xylulose reductase of filamentous fungi identified in Aspergillus niger.
PMID 20654618
Abstract
L-Xylulose reductase is part of the eukaryotic pathway for l-arabinose catabolism. A previously identified L-xylulose reductase in Hypocrea jecorina turned out to be not the 'true' one since it was not upregulated during growth on L-arabinose and the deletion strain showed no reduced L-xylulose reductase activity but instead lost the D-mannitol dehydrogenase activity. In this communication we identified the 'TRUE'L-xylulose reductase in Aspergillus niger. The gene, lxrA (JGI177736), is upregulated on L-arabinose and the deletion results in a strain lacking the NADPH-specific L-xylulose reductase activity and having reduced growth on l-arabinose. The purified enzyme had a K(m) for L-xylulose of 25 mM and a nu(max) of 650 U/mg.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
68854
L-Arabino-1,4-lactone, ≥95.0% (GC)
C5H8O5