EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Beta-glucuronidase from Helix pomatia origin is not suitable for diosmetin analysis.
PMID 20656432
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7321
Diosmetin
C16H12O6
90985
Diosmetin, analytical standard
C16H12O6