EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Solid-phase synthetic route to multiple derivatives of a fundamental peptide unit.
PMID 20657403
Abstract
Amino acids are Nature's combinatorial building blocks. When substituted on both the amino and carboxyl sides they become the basic scaffold present in all peptides and proteins. We report a solid-phase synthetic route to large combinatorial variations of this fundamental scaffold, extending the variety of substituted biomimetic molecules available to successfully implement the Distributed Drug Discovery (D3) project. In a single solid-phase sequence, compatible with basic amine substituents, three-point variation is performed at the amino acid a-carbon and the amino and carboxyl functionalities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
123889
α,α′-Dichloro-o-xylene, 98%
C8H8Cl2