EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Synthesis of 2-(2-aminopyrimidine)-2,2-difluoroethanols as potential bioisosters of salicylidene acylhydrazides.
PMID 20657451
Abstract
Salicylidene acylhydrazides are inhibitors of type III secretion in several gram-negative pathogens. To further develop the salicylidene acylhydrazides, scaffold hopping was applied to replace the core fragment of the compounds. The novel 2-(2-amino-pyrimidine)-2,2-difluoroethanol scaffold was identified as a possible analog to the salicylidene acylhydrazide core structure. The synthesis of a library of 2-(2-amino-pyrimidine)-2,2-difluoro-ethanols is described in this paper.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A78608
2-Aminopyrimidine, 97%
C4H5N3