EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
The reaction of cyanoacetylhydrazine with omega-bromo(4-methyl)acetophenone: synthesis of heterocyclic derivatives with antitumor activity.
PMID 20657502
Abstract
New approaches for the synthesis of hydrazide-hydrazone derivatives were demonstrated as well as some heterocyclizations of such derivatives to afford 1,3,4-triazine, pyridine and 1,3,4-oxadiazine derivatives. The antitumor evaluation of the newly synthesized products against three cancer cell lines, namely breast adenocarcinoma (MCF-7), non-small cell lung cancer (NCI-H460) and CNS cancer (SF-268) were recorded. Most of the synthesized compounds showed high inhibitory effects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8309
Acethydrazide, 90%
C2H6N2O