EMAIL THIS PAGE TO A FRIEND
American journal of kidney diseases : the official journal of the National Kidney Foundation
Corosolic acid-induced acute kidney injury and lactic acidosis in a patient with impaired kidney function.
PMID 20659629
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G0549
Corosolic acid from Lagerstroemia speciosa, ≥98% (HPLC)
C30H48O4