EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Palladium-catalyzed coupling of aldehyde-derived hydrazones: practical synthesis of triazolopyridines and related heterocycles.
PMID 20661974
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
697230
[1,1′-Bis(diphenylphosphino)ferrocene]dichloropalladium(II)
C34H28Cl2FeP2Pd