EMAIL THIS PAGE TO A FRIEND
Journal of pediatric endocrinology & metabolism : JPEM
Chronic intermittent form of isovaleric acidemia mimicking diabetic ketoacidosis.
PMID 20662350
Abstract
Isovaleric acidemia is a rare autosomal recessive inborn error of leucine metabolism. Two phenotypes with either an acute neonatal or a chronic intermittent presentation were described. The acute type is observed more frequently and is more fatal. We report the case of a girl in childhood who presented with hyperglycemia and metabolic acidosis with an increased anion gap; and preliminarily diagnosed as diabetic ketoacidosis, but further investigation revealed chronic intermittent isovaleric academia. This case is of interest because of the rarity of this presentation. The importance of thinking for inborn errors of metabolism in children with metabolic acidosis in late childhood is emphasized.
Related Materials