EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Mesoporous beta-Ni(OH)2: synthesis and enhanced electrochemical performance.
PMID 20664885
Abstract
Mesoporous beta-Ni(OH)(2) synthesized by a soft template method as a cathode material in a nickel based battery can provide higher specific discharge capacity in comparison with other micro/nanostructures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
283622
Nickel(II) hydroxide
H2NiO2