EMAIL THIS PAGE TO A FRIEND
Biopolymers
Enzymatic activity and thermal stability of metallo proteins in hydrated ionic liquids.
PMID 20665687
Abstract
Hydrated choline dihydrogen phosphate (Hy[ch][dhp]) containing 30 wt% water was investigated as a novel protein solvent. The Hy[ch][dhp] dissolved some metallo proteins (cytochrome c, peroxidase, ascorbate oxidase, azurin, pseudoazurin and fructose dehydrogenase) without any modification. These proteins retained the surroundings of the active site after dissolution in Hy[ch][dhp]. Some metallo proteins were found to retain their activity in the Hy[ch][dhp].
Related Materials