EMAIL THIS PAGE TO A FRIEND
Acta Physiologica (Oxford, England)
Opposing responses of apoptosis and autophagy to moderate compression in skeletal muscle.
PMID 20670304
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-702
Anti-FOXO3a/FKHRL1 Antibody, clone, Upstate®, from rabbit