EMAIL THIS PAGE TO A FRIEND
Neuron
Basal GABA regulates GABA(B)R conformation and release probability at single hippocampal synapses.
PMID 20670833
Abstract
Presynaptic GABA(B) receptor (GABA(B)R) heterodimers are composed of GB(1a)/GB(2) subunits and critically influence synaptic and cognitive functions. Here, we explored local GABA(B)R activation by integrating optical tools for monitoring receptor conformation and synaptic vesicle release at individual presynaptic boutons of hippocampal neurons. Utilizing fluorescence resonance energy transfer (FRET) spectroscopy, we detected a wide range of FRET values for CFP/YFP-tagged GB(1a)/GB(2) receptors that negatively correlated with release probabilities at single synapses. High FRET of GABA(B)Rs associated with low release probability. Notably, pharmacological manipulations that either reduced or increased basal receptor activation decreased intersynapse variability of GB(1a)/GB(2) receptor conformation. Despite variability along axons, presynaptic GABA(B)R tone was dendrite specific, having a greater impact on synapses at highly innervated proximal branches. Prolonged neuronal inactivity reduced basal receptor activation, leading to homeostatic augmentation of release probability. Our findings suggest that local variations in basal GABA concentration are a major determinant of GB(1a)/GB(2) conformational variability, which contributes to heterogeneity of neurotransmitter release at hippocampal synapses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S9066
SKF-89976A, >98% (HPLC), solid
C22H25NO2 · HCl