EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
The phosphatase interactor NIPP1 regulates the occupancy of the histone methyltransferase EZH2 at Polycomb targets.
PMID 20671031
Abstract
Polycomb group (PcG) proteins are key regulators of stem-cell and cancer biology. They mainly act as repressors of differentiation and tumor-suppressor genes. One key silencing step involves the trimethylation of histone H3 on Lys27 (H3K27) by EZH2, a core component of the Polycomb Repressive Complex 2 (PRC2). The mechanism underlying the initial recruitment of mammalian PRC2 complexes is not well understood. Here, we show that NIPP1, a regulator of protein Ser/Thr phosphatase-1 (PP1), forms a complex with PP1 and PRC2 components on chromatin. The knockdown of NIPP1 or PP1 reduced the association of EZH2 with a subset of its target genes, whereas the overexpression of NIPP1 resulted in a retargeting of EZH2 from fully repressed to partially active PcG targets. However, the expression of a PP1-binding mutant of NIPP1 (NIPP1m) did not cause a redistribution of EZH2. Moreover, mapping of the chromatin binding sites with the DamID technique revealed that NIPP1 was associated with multiple PcG target genes, including the Homeobox A cluster, whereas NIPP1m showed a deficient binding at these loci. We propose that NIPP1 associates with a subset of PcG targets in a PP1-dependent manner and thereby contributes to the recruitment of the PRC2 complex.