EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and evaluation of oryzalin analogs against Toxoplasma gondii.
PMID 20675138
Abstract
The synthesis and evaluation of 20 dinitroanilines and related compounds against the obligate intracellular parasite Toxoplasma gondii is reported. Using in vitro cultures of parasites in human fibroblasts, we determined that most of these compounds selectively disrupted Toxoplasma microtubules, and several displayed sub-micromolar potency against the parasite. The most potent compound was N(1),N(1)-dipropyl-2,6-dinitro-4-(trifluoromethyl)-1,3-benzenediamine (18b), which displayed an IC(50) value of 36 nM against intracellular T. gondii. Based on these data and another recent report [Ma, C.; Tran, J.; Gu, F.; Ochoa, R.; Li, C.; Sept, D.; Werbovetz, K.; Morrissette, N. Antimicrob. Agents Chemother. 2010, 54, 1453], an antimitotic structure-activity relationship for dinitroanilines versus Toxoplasma is presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36182
Oryzalin, PESTANAL®, analytical standard
C12H18N4O6S
N12729
Oryzalin, analytical standard
C12H18N4O6S