EMAIL THIS PAGE TO A FRIEND
Archives of disease in childhood. Education and practice edition
Sedation for radiological imaging.
PMID 20675522
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T8275
Temazepam, analytical standard
C16H13ClN2O2