EMAIL THIS PAGE TO A FRIEND
Toxicology letters
The role of cyclooxygenase-2-dependent signaling via cyclic AMP response element activation on aromatase up-regulation by o,p'-DDT in human breast cancer cells.
PMID 20678559
Abstract
o,p'-Dichlorodiphenyltrichloroethane (o,p'-DDT) is a DDT isomer and xenoestrogen that can induce inflammation and cancer. However, the effect of o,p'-DDT on aromatase is unclear. Thus, we investigated the effects of o,p'-DDT on aromatase expression in human breast cancer cells. We also examined whether cyclooxygenase-2 (COX-2) is involved in o,p'-DDT-mediated aromatase expression. Treatment with o,p'-DDT-induced aromatase protein expression in MCF-7 and MDA-MB-231 human breast cancer cells; enhancing aromatase gene expression, and enzyme and promoter activity. Treatment with ICI 182.780, a estrogen receptor antagonist, did not affect the inductive effects of o,p'-DDT on aromatase expression. In addition, o,p'-DDT increased COX-2 protein levels markedly, increased COX-2 mRNA expression and promoter activity, enhanced the production of prostaglandin E(2) (PGE(2)), induced cyclic AMP response element (CRE) activation, and cAMP levels and binding of CREB. o,p'-DDT also increased the phosphorylation of PKA, Akt, ERK, and JNK in their signaling pathways in MCF-7 and MDA-MB-231 cells. Finally, o,p'-DDT induction of aromatase was inhibited by various inhibitors [COX-2 (by NS-398), PKA (H-89), PI3-K/Akt (LY 294002), EP2 (AH6809), and EP4 receptor (AH23848)]. Together, these results suggest that o,p'-DDT increases aromatase, and that o,p'-DDT-induced aromatase is correlated with COX-2 up-regulation, mediated via the CRE activation and PKA and PI3-kinase/Akt signaling pathways in breast cancer cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N12708 2,4′-DDT, analytical standard
C14H9Cl5
A8227
AH23848 hemicalcium salt, ≥90% (HPLC), powder
C29H34NO5 · 0.5 Ca