EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Lactones and quinones from the tubers of Sinningia aggregata.
PMID 20684540
Abstract
Three new aromatic epsilon-lactones, aggregatins A (1), B (2), and C (3), a new naphthoquinone derivative, aggregatin D (4), and three known anthraquinones, 2-methylanthraquinone, 7-methoxy-2-methylanthraquinone, and 7-hydroxy-2-methylanthraquinone, were isolated from the tubers of Sinningia aggregata (Gesneriaceae). Compounds 1 and 4 and the anthraquinones showed marginal antimicrobial activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
65800
2-Methylanthraquinone, technical, ≥95% (HPLC)
C15H10O2