EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Biomimetic photocycloaddition of 3-hydroxyflavones: synthesis and evaluation of rocaglate derivatives as inhibitors of eukaryotic translation.
PMID 20687060