EMAIL THIS PAGE TO A FRIEND
The Cochrane database of systematic reviews
Antibiotic therapy for Shigella dysentery.
PMID 20687081
Abstract
Shigella dysentery is a relatively common illness and occasionally causes death, worldwide. Mild symptoms are self-limiting but in more severe cases, antibiotics are recommended for cure and preventing relapse. The antibiotics recommended are diverse, have regional differences in sensitivity, and have side effects. To evaluate the efficacy and safety of antibiotics for treating Shigella dysentery. In June 2009 we identified all relevant trials from the following databases: Cochrane Infectious Diseases Group Specialized Register; Cochrane Central Register of Controlled Trials (CENTRAL) (The Cochrane Library 2008, issue 4), MEDLINE, EMBASE, LILACS and the metaRegister of Controlled Trials (mRCT). We also checked conference proceedings for relevant abstracts, and contacted researchers, organizations, and pharmaceutical companies. Randomized controlled trials of antibiotics for Shigella dysentery. Four authors, working in pairs, independently assessed trial eligibility, methodological quality, and extracted data. We calculated risk ratios (RR) with 95% confidence intervals (CI) for dichotomous data, and used the random-effects model for significant heterogeneity. We explored possible sources of heterogeneity, when present, in subgroup analyses of participant age and percentage of participants with confirmed Shigella infection. Sixteen trials (1748 participants), spanning four decades and with differing sensitivity to Shigella isolates, met the inclusion criteria. Seven were judged to be at risk of bias due to inadequate allocation concealment or blinding, and 12 due to incomplete reporting of outcome data. Limited data from one three-armed trial of people with moderately severe illness suggest that antibiotics reduce the episodes of diarrhoea at follow-up (furazolidone versus no drug RR 0.21, 95% CI 0.09 to 0.48, 73 participants; cotrimoxazole versus no drug RR 0.30, 95% CI 0.15 to 0.59, 76 participants).There was insufficient evidence to consider any class of antibiotic superior in efficacy in treating Shigella dysentery, but heterogeneity for some comparisons limits confidence in the results. All the antibiotics studied were safe. There was inadequate evidence regarding the role of antibiotics in preventing relapses. Antibiotics reduce the duration of Shigella dysentery.Regularly updated local or regional antibiotic sensitivity patterns to different species and strains of Shigella are required to guide empiric therapy. More trials adhering to standard guidelines are required to evaluate the role of antibiotics in the treatment of severe forms of Shigella dysentery and in groups who are at high risk of complications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F9505
Furazolidone
C8H7N3O5
46297
Furazolidone, VETRANAL, analytical standard
C8H7N3O5