EMAIL THIS PAGE TO A FRIEND
Molecular cancer
MiR-145, a new regulator of the DNA fragmentation factor-45 (DFF45)-mediated apoptotic network.
PMID 20687965
Abstract
MicroRNA-145 (miR-145) is considered to play key roles in many cellular processes, such as proliferation, differentiation and apoptosis, by inhibiting target gene expression. DNA Fragmentation Factor-45 (DFF45) has been found to be the substrate of Caspase-3, and the cleavage of DFF45 by caspase-3 during apoptosis releases DFF40 that degrades chromosomal DNA into nucleosomal fragments. There are currently no in-depth studies on the relationship between miR-145 and the DFF45 gene. In this study, we identified DFF45 as a novel target of miR-145. We demonstrated that miR-145 targets a putative binding site in the coding sequence (CDS) of DFF45, and its abundance is inversely associated with DFF45 expression in colon cancer cells. Using a luciferase reporter system, we found that miR-145 suppresses the expression of the luciferase reporter gene fused to the putative binding site of DFF45. The level of DFF45 protein, but not DFF45 mRNA, was decreased by miR-145, suggesting a mechanism of translational regulation. Furthermore, we demonstrate that this specific silencing of DFF45 by miR-145 accounts, at least in part, for the staurosporine-induced tumor cell apoptosis in vitro. Our study reveals a previously unrecognized function of miR-145 in DFF45 processing, which may underlie crucial aspects of cancer biology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405710
Anti-DFFA antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV30359
Anti-DFFA antibody produced in rabbit, IgG fraction of antiserum
HPA025230
Anti-DFFA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3
HPA018859
Anti-DFFA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA019938
Anti-DFFA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB4500644
Anti-DFFA antibody produced in rabbit, affinity isolated antibody
PRS2001
Anti-ICAD antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS2003
Anti-ICAD antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
CKOZFN7161 CompoZr® Knockout ZFN Kit, Human DFFA (NM_004401)
CKOZFND7161 CompoZr® Knockout ZFN Kit, ZFN plasmid only, Human DFFA (NM_004401)
SAB4200199
Monoclonal Anti-DFF45 antibody produced in mouse, ~1.0 mg/mL, clone 3F12, purified immunoglobulin
SAB4100013
Monoclonal Anti-DFFA antibody produced in mouse, culture supernatant
SAB1402172
Monoclonal Anti-DFFA, (C-terminal) antibody produced in mouse, clone 3A11, purified immunoglobulin, buffered aqueous solution