American journal of physiology. Gastrointestinal and liver physiology
Nuclear expression of interleukin-33 in pancreatic stellate cells.
PMID 20689058
Activated pancreatic stellate cells (PSCs) play a pivotal role in pancreatic fibrosis in chronic pancreatitis and pancreatic cancer. Recent studies have suggested a role of IL-33, a newly identified IL-1 family member, in fibrosis. We here examined the expression of IL-33 and the IL-33-mediated regulation of cell functions in PSCs. PSCs were isolated from human and rat pancreas tissues. The expression of IL-33 was examined by Western blotting, PCR, ELISA, and immunostaining. The roles of IL-33 in the regulation of PSC functions were examined by using recombinant IL-33 and small interfering RNA. Activated PSCs expressed IL-33 in the nucleus, and the expression was increased by IL-1β, TNF-α, PDGF-BB, and IFN-γ, but not TGF-β1. Nuclear IL-33 expression was also observed in the pancreatic acinar and ductal cells. IL-1β induced IL-33 expression mainly through the activation of NF-κB and ERK pathways and partially through that of p38 MAP kinase, whereas PDGF-BB induced IL-33 expression mainly through the activation of ERK pathway. PSCs expressed soluble ST2, ST2L, and IL-1RAcP, but the expression level of ST2L was relatively low. Recombinant IL-33 did not stimulate key cell functions of PSCs. Decreased IL-33 expression by small interfering RNA resulted in decreased proliferation in response to PDGF-BB. In conclusion, activated PSCs expressed IL-33 in the nucleus. IL-33 might regulate the PDGF-induced proliferation in PSCs.