EMAIL THIS PAGE TO A FRIEND
Tetrahedron letters
Microwave-assisted Organic Synthesis of a High-affinity Pyrazolo-pyrimidinyl TSPO Ligand.
PMID 20689673
Abstract
We herein report a dramatically improved total synthesis of the high-affinity translocator protein (TSPO) ligand DPA-714, featuring microwave-assisted organic synthesis (MAOS). Compared with previously described approaches, our novel MAOS method dramatically reduces overall reaction time without adversely effecting reaction yields. We envision that the described MAOS protocol may be suitably applied to high-throughput, diversity-oriented synthesis of novel compounds based on the pyrazolo-pyrimidinyl scaffold. Such an approach could accelerate the development of focused libraries of novel TSPO ligands with potential for future development as molecular imaging and therapeutic agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
250996
2-Chloro-N,N-diethylacetamide, 97%
C6H12ClNO