EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Synthesis of indolequinones from bromoquinones and enamines mediated by Cu(OAc)2.H2O.
PMID 20690687
Abstract
A Cu(II)-mediated synthesis of indolequinones from the corresponding bromoquinones and enamines is reported. The key oxidative cyclization proceeds in good yield for a broad range of substrates and can be performed on a multigram scale, allowing access to biologically interesting structures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
517453
Copper(II) acetate, powder, 99.99% trace metals basis
C4H6CuO4
326755
Copper(II) acetate, 98%
C4H6CuO4