EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Discussion on "A comprehensive two-dimensional gas chromatography coupled with quadrupole mass spectrometry approach for identification of C10 derivatives from decalin" by C. Flego, N. Gigantiello, W.O. Parker, Jr., V. Calemma [J. Chromatogr. A 1216 (2009) 2891].
PMID 20696435