EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Main-group and transition-element IRMOF homologues.
PMID 20698648
Abstract
A simple two-component procedure was developed to synthesize not only classical zinc-based IRMOFs represented by MOF-5 but also the cobalt and beryllium homologues of this most prominent MOF. The procedure is the first manifestation of mirroring the IRMOF series with other metal centers taken from main-group as well as transition-metal elements. Because of the existence of many suitable precursors, the procedure promises the generation of a large number of IRMOF homologues. Since the IRMOF series together with the MIL series is the MOF group with the largest number of representatives, the possibility of choosing the metal centers of the secondary building units from an extended set will tremendously expand the number of obtainable structures in a predictive, crystal-engineering-type way. Use of metal centers other than zinc will allow the addition of new features to the existing IRMOF structures, such as magnetic properties in the example of cobalt.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
749125
Cobalt(II) oxo pivalate
C60H108Co8O26
749141
Iron(III) oxo acetate perchlorate hydrate
C12H24Fe3O16 · ClO4 · xH2O
749168
Zirconium(IV) oxo hydroxy methacrylate
C48H64O32Zr6