EMAIL THIS PAGE TO A FRIEND
Acta tropica
Antioxidant, larvicidal and antiacetylcholinesterase activities of cashew nut shell liquid constituents.
PMID 20707981
Abstract
Anacardic acid, cardanol and cardol, the main constituents of natural cashew nut shell liquid (CNSL), were obtained by solvent extraction and assayed for antioxidant, larvicidal and antiacetylcholinesterase activity. Their relative percent composition was obtained by HPLC analysis. Antioxidant activity was assessed using the DPPH and ABTS(+) tests, which showed cardanol as the most active, followed by cardol and anacardic acid. The three CNSL components demonstrated good larvicidal activity against Aedes aegypti (LC(50)=12.40 for anacardic acid, 10.22 for cardol and 14.45 for cardanol) and exhibited inhibition zones for acetylcholinesterase enzymes in the TLC test similar to carbachol, which was used as standard. Based on the results, these multipotent compounds represent promising agents in the control of Ae. aegypti, the main dengue vector in Brazil.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
91822
5-Pentadecylresorcinol, analytical standard
C21H36O2