EMAIL THIS PAGE TO A FRIEND
Biomolecular NMR assignments
Backbone chemical shifts assignments of D-allose binding protein in the free form and in complex with D-allose.
PMID 20711759
Abstract
D-allose binding protein (ALBP) belongs to the family of perisplamic receptors of the bacterial ABC transporter system. ALBP experiences a significant conformational rearrangement upon binding to the sugar. Here, we report the sequential backbone assignment for the ALBP from Escherichia coli in the free form (BMRB no. 16982) and in complex with D-allose (BMRB no. 16984).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
285005
D-Allose, 98%
C6H12O6