EMAIL THIS PAGE TO A FRIEND
Journal of molecular modeling
Mechanistic investigation of the base-promoted cycloselenoetherification of pent-4-en-1-ol.
PMID 20711793
Abstract
The mechanism of phenylselenoetherification of pent-4-en-1-ol using some bases (pyridine, triethylamine, quinoline, 2,2'-bipyridine) as catalyst was examined through studies of kinetics of the cyclization, by UV-VIS spectrophotometry. It was demonstrated that the intramolecular cyclization is facilitated in the presence of bases caused by the hydrogen bond between base and alkenol's OH-group. The obtained values for rate constants have shown that the reaction with triethylamine is the fastest one. Quantum chemical calculations (MP2(fc)/6-311+G**//B3LYP/6-311+G**) show, that the transition state of the cyclisation is S(N)2 like.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
111279
4-Penten-1-ol, 99%
C5H10O