EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Cobalt-catalyzed hydroarylation of alkynes through chelation-assisted C-H bond activation.
PMID 20712366
Abstract
Ternary catalytic systems consisting of cobalt salts, phosphine ligands, and Grignard reagents promote addition of arylpyridines and imines to unactivated internal alkynes with high regio- and stereoselectivities. Deuterium-labeling experiments suggest that the reaction involves chelation-assisted oxidative addition of the aryl C-H bond to the cobalt center and insertion of the C-C triple bond into the Co-H bond, followed by reductive elimination of the resulting diorganocobalt species.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P33402
2-Phenylpyridine, 98%
C11H9N