EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Synthesis of a mesoporous single crystal Ga2O3 nanoplate with improved photoluminescence and high sensitivity in detecting CO.
PMID 20714548
Abstract
A new approach is proposed to synthesize a mesoporous single crystal Ga(2)O(3) nanoplate by heating a single crystal nanoplate of GaOOH, which involves an ion exchange between KGaO(2) and CH(3)COOH at room temperature for the formation of GaOOH and pseudomorphic and topotactic phase transformation from GaOOH to Ga(2)O(3).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
215066
Gallium(III) oxide, ≥99.99% trace metals basis
Ga2O3