EMAIL THIS PAGE TO A FRIEND
Seikagaku. The Journal of Japanese Biochemical Society
[Functions and biosynthesis pathway of sulfur-modifications in tRNA].
PMID 20715575
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T4509
4-Thiouridine, ≥98%
C9H12N2O5S