EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Cyclometalated red iridium(III) complexes containing carbazolyl-acetylacetonate ligands: efficiency enhancement in polymer LED devices.
PMID 20717556
Abstract
The design, synthesis, photophysical and significantly improved electrooptical properties of a series of red emitting cyclometalated iridium(iii) complexes containing carbazolyl-acetylacetonate ligands are described.