EMAIL THIS PAGE TO A FRIEND
Organic letters
Diamine-free lithiation-trapping of N-Boc heterocycles using s-BuLi in THF.
PMID 20718476
Abstract
A diamine-free protocol for the s-BuLi-mediated lithiation-trapping of N-Boc heterocycles has been developed. In the optimized procedure, lithiation is accomplished using s-BuLi in THF at -30 °C for only 5 or 10 min. Subsequent electrophilic trapping or transmetalation-Negishi coupling delivered a range of functionalized pyrrolidines, imidazolidines, and piperazines in 43-83% yield.