EMAIL THIS PAGE TO A FRIEND
Nature reviews. Endocrinology
Diabetes: Treatment of diabetes mellitus: new tricks by an old player.
PMID 20720590
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0070
Salsalate, ≥98% (HPLC)
C14H10O5