EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Discovery of quinolines as selective glucocorticoid receptor agonists.
PMID 20727743
Abstract
The dissociated glucocorticoid receptor (GR) agonist ZK 216348 is rendered GR-selective over other nuclear hormone receptors through replacing the methylbenzoxazine with a quinoline moiety. Compounds were shown to be efficacious in cell assays with respect to inflammation endpoints, along with reduced activity in a transactivation assay, hinting at an improved therapeutic window over corticosteroids.