EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Optimization of N-benzyl-benzoxazol-2-ones as receptor antagonists of macrophage migration inhibitory factor (MIF).
PMID 20728358
Abstract
The cytokine MIF is involved in inflammation and cell proliferation via pathways initiated by its binding to the transmembrane receptor CD74. MIF also exhibits keto-enol tautomerase activity, believed to be vestigial in mammals. Starting from a 1 μM hit from virtual screening, substituted benzoxazol-2-ones have been discovered as antagonists with IC(50) values as low as 7.5 nM in a tautomerase assay and 80 nM in a MIF-CD74 binding assay. Additional studies for one of the potent inhibitors demonstrated that it is not a covalent inhibitor of MIF and that it attenuates MIF-dependent ERK1/2 phosphorylation in human synovial fibroblasts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
157058
2-Benzoxazolinone, 98%
C7H5NO2