EMAIL THIS PAGE TO A FRIEND
Biological psychiatry
Effects of an alpha 7-nicotinic agonist on default network activity in schizophrenia.
PMID 20728875
Abstract
3-(2,4-dimethoxybenzylidene)-anabaseine (DMXB-A) is a partial agonist at α7 nicotinic acetylcholine receptors that has been evaluated clinically for treatment of schizophrenia. This study examined the effects of DMXB-A on default network activity as a biomarker for drug effects on pathologic brain function associated with schizophrenia. Placebo and two doses of DMXB-A were administered in a random, double-blind crossover design during a Phase 2 study of DMXB-A. Functional magnetic resonance imaging was performed on 16 nonsmoking patients with schizophrenia while they performed a simple eye movement task. Independent component analysis was used to identify the default network component. Default network changes were evaluated in the context of a polymorphism in CHRNA7, the α7-nicotinic acetylcholine receptor subunit gene, which was previously found to be associated with schizophrenia. Compared with placebo, both 150 and 75 mg twice daily DMXB-A altered default network activity, including a reduction in posterior cingulate, inferior parietal cortex, and medial frontal gyrus activity and an increase in precuneus activity. The most robust difference, posterior cingulate activity reduction, was affected by CHRNA7 genotype. The observed DMXB-A-related changes are consistent with improved default network function in schizophrenia. Pharmacogenetic analysis indicates mediation of the effect through the α7-nicotinic receptor. These results further implicate nicotinic cholinergic dysfunction in the disease and suggest that default network activity may be a useful indicator of biological effects of novel therapeutic agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0326
GTS-21, ≥97% (HPLC)
C19H20N2O2 · 2HCl