EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Electrochemical genotyping and detection of single-nucleotide polymorphisms based on junction-probe containing 2'-deoxyinosine.
PMID 20730202
Abstract
A novel electrochemical biosensor for the genotyping/detection of single-nucleotide polymorphisms (SNPs) of trace oral-cancer salivary DNA based on junction probes containing 2'-deoxyinosine (dI) residues is described.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5287
2′-Deoxyinosine, ≥98%
C10H12N4O4