EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Controlled production of polymer microspheres from microgel-stabilized high internal phase emulsions.
PMID 20730203
Abstract
We describe a flexible method for large-scale production of polymeric microspheres by templating a microgel-stabilized oil-in-water (o/w) high internal phase emulsion.