EMAIL THIS PAGE TO A FRIEND
Natural product reports
Trends for diverse production strategies of plant medicinal alkaloids.
PMID 20730220
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
41111
Ajmalicine, ≥98.0% (HPLC)
C21H24N2O3