EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
High-throughput analysis of hexosamine using a colorimetric method.
PMID 20732290
Abstract
A 96-well plate method was developed for analysis of total hexosamine content in biological samples. Four hexosamine monomer derivatives-glucosamine hydrochloride, glucosamine sulfate, galactosamine hydrochloride, and mannosamine hydrochloride-were examined for the linearity of their spectra in the concentration range specified in the assay. The hexosamine concentration analysis range was linear from 0.1 to 1 mM. The quantification of hexosamines from chitin and chitosan upon acid hydrolysis was also tested. Accurate quantification of glucosamine content in chitin and chitosan with different molecular sizes and degrees of acetylation was demonstrated using the new method.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M4670
D-Mannosamine hydrochloride
C6H13NO5 · HCl